2,3,6-Trichlorobenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 2,3,6-TBA, 2,3,6-trichlorobenzoic acid
T3758-1G T3758-5G
TCIAT3758-1GEA 660 NOK
TCIAT3758-1G TCIAT3758-5G
2,3,6-Trichlorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3,6-Trichlorobenzoic acid
Formel: C₇H₃Cl₃O₂
MW: 225,46 g/mol
MDL nummer: MFCD00053310
CAS-nummer: 50-31-7
EINECS: 200-026-4
FN: 3077
ADR: 9,III
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 130.0 deg-C