Trimethylsulphoxonium chloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Trimethyloxosulphonium chloride
T3794-25G
T3794-5G
TCIAT3794-25GEA
3625
NOK
TCIAT3794-25G
TCIAT3794-5G
Trimethylsulphoxonium chloride ≥98.0% (by titrimetric analysis)
Trimethylsulphoxonium chloride
Formel:
C₃H₉ClOS MW: 128,62 g/mol |
MDL nummer:
MFCD00011898 CAS-nummer: 5034-06-0 EINECS: 225-724-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 240.0 to 242.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...