(Triethoxysilyl)methyl methacrylate ≥97.0% (by GC) stabilised
Leverandør: TCI
|
Warning
|
Synonymer:
Methacrylic acid (Triethoxysilyl)methyl ester
Stabilisering:
Stabilised with 4-methoxyphenol (MEHQ)
T3852-5G
T3852-25G
TCIAT3852-5GEA
595
NOK
TCIAT3852-5G
TCIAT3852-25G
(Triethoxysilyl)methyl methacrylate ≥97.0% (by GC) stabilised
(Triethoxysilyl)methyl methacrylate
Formel:
C₁₁H₂₂O₅Si MW: 262,38 g/mol Kokepunkt: 97°C |
MDL nummer:
MFCD00156470 CAS-nummer: 5577-72-0 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...