Tizoxanide ≥95.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 2-Hydroxy-N-(5-nitrothiazol-2-yl)benzamide, 2-Hydroxy-N-(5-nitro-2-thiazolyl)benzamide

T3862-100MG T3862-25MG
TCIAT3862-100MGEA 1750 NOK
TCIAT3862-100MG TCIAT3862-25MG
Tizoxanide ≥95.0% (by HPLC)
Tizoxanide
Formel: C₁₀H₇N₃O₄S
MW: 265,24 g/mol
Smeltepunkt: 254°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD07484970
CAS-nummer: 173903-47-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 15.10 to 16.60 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR