Triethoxy(2,4,4-trimethylpentyl)silane ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Isooctyltriethoxysilane
T3883-100ML
T3883-25ML
TCIAT3883-100MLEA
1025
NOK
TCIAT3883-100ML
TCIAT3883-25ML
Triethoxy(2,4,4-trimethylpentyl)silane ≥97.0% (by GC)
Triethoxy(2,4,4-trimethylpentyl)silane
Formel:
C₁₄H₃₂O₃Si MW: 276,49 g/mol Flash punkt: 42°C |
MDL nummer:
MFCD09838917 CAS-nummer: 35435-21-3 EINECS: 252-558-1 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...