Uramil-N,N-diacetic acid monohydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

U0014-1G U0014-5G
TCIAU0014-1GEA 620 NOK
TCIAU0014-1G TCIAU0014-5G
Uramil-N,N-diacetic acid monohydrate ≥98.0% (by titrimetric analysis)
Uramil-N,N-diacetic acid monohydrate
Formel: C₈H₉N₃O₇·H₂O
MDL nummer: MFCD00047556
CAS-nummer: 13055-06-6
EINECS: 235-941-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) min. 98.0 % (as 1H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR