Valeraldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Pentanal 2,4-Dinitrophenylhydrazone
V0066-1G
V0066-10G
TCIAV0066-1GEA
300
NOK
TCIAV0066-1G
TCIAV0066-10G
Valeraldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Valeraldehyde 2,4-Dinitrophenylhydrazone
Formel:
C₁₁H₁₄N₄O₄ MW: 266,26 g/mol |
MDL nummer:
MFCD00191595 CAS-nummer: 2057-84-3 FN: 1325 ADR: 4.1,III |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 108.0 to 111.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...