4-Vinyl-1,3-dioxolan-2-one ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
V0114-25G
V0114-100G
TCIAV0114-25GEA
500
NOK
TCIAV0114-25G
TCIAV0114-100G
4-Vinyl-1,3-dioxolan-2-one ≥98.0% (by GC)
4-Vinyl-1,3-dioxolan-2-one
Formel:
C₅H₆O₃ MW: 114,10 g/mol Kokepunkt: 122°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00143315 CAS-nummer: 4427-96-7 FN: 2810 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...