L-Valine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: H-Val-OEt·HCl, (S)-Ethyl 2-amino-3-methylbutanoate hydrochloride

V0142-5G V0142-25G
TCIAV0142-5GEA 270 NOK
TCIAV0142-5G TCIAV0142-25G
L-Valine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-Valine ethyl ester hydrochloride
Formel: C₇H₁₆ClNO₂
MW: 181,66 g/mol
Smeltepunkt: 104°C
MDL nummer: MFCD00012511
CAS-nummer: 17609-47-1
EINECS: 241-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e102591de56a37d4792ad539aae0a42a6194a2b&vid=5e102591de56a37d4792ad539aae0a42a6194a2b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +6.0 to +8.0 deg (C=2,H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR