VX-702 ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 2-(2,4-Difluorophenyl)-6-(1-(2,6-difluorophenyl)ureido)nicotinamide, 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide

V0147-25MG V0147-100MG
TCIAV0147-25MGEA 1875 NOK
TCIAV0147-25MG TCIAV0147-100MG
VX-702 ≥98.0% (by HPLC)
VX-702
Formel: C₁₉H₁₂F₄N₄O₂
MW: 404,32 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD11616590
CAS-nummer: 745833-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59384aef51ddf793b60f592f0271e8114c51d49f&vid=59384aef51ddf793b60f592f0271e8114c51d49f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 13.00 to 14.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR