WAY-100635 Maleate ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
WAY-100635 maleate, N-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinyl-cyclohexanecarboxamide 2Z-butenedioate, N-[2-[4-(2-Methoxyphenyl)piperazin-1-yl]ethyl]-N-(pyridin-2-yl)cyclohexanecarboxamide maleate
W0016-10MG
W0016-50MG
TCIAW0016-10MGEA
2175
NOK
TCIAW0016-10MG
TCIAW0016-50MG
WAY-100635 Maleate ≥98.0% (by HPLC)
WAY-100635 Maleate
Formel:
C₂₅H₃₄N₄O₂·C₄H₄O₄ MW: 538,65 g/mol |
MDL nummer:
MFCD01321056 CAS-nummer: 1092679-51-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...