Xylobiose ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 4-O-beta-D-Xylopyranosyl-D-xylose

X0067-100MG X0067-1G
TCIAX0067-100MGEA 1475 NOK
TCIAX0067-100MG TCIAX0067-1G
Xylobiose ≥98.0% (by HPLC)
Xylobiose
Formel: C₁₀H₁₈O₉
MW: 282,25 g/mol
MDL nummer: MFCD00135984
CAS-nummer: 6860-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=001e8e99e51b8b83bd805fb59afa83b78670196b&vid=001e8e99e51b8b83bd805fb59afa83b78670196b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -24.0 to -28.0 deg (C=10、H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR