m-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

X0076-200MG X0076-1G
TCIAX0076-200MGEA 795 NOK
TCIAX0076-200MG TCIAX0076-1G
m-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)
m-Xylylenediphosphonic acid
Formel: C₈H₁₂O₆P₂
MW: 266,13 g/mol
MDL nummer: MFCD22200402
CAS-nummer: 144052-40-4
FN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e37d211d02f1b3cde231c79e3730c37ac767f42&vid=9e37d211d02f1b3cde231c79e3730c37ac767f42&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR