Ziprasidone ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: Ziprasidonum, 5-(2-(4-(Benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindolin-2-one

Z0034-100MG Z0034-1G
TCIAZ0034-100MGEA 865 NOK
TCIAZ0034-100MG TCIAZ0034-1G
Ziprasidone ≥98.0% (by HPLC)
Ziprasidone
Formel: C₂₁H₂₁ClN₄OS
MW: 412,94 g/mol
MDL nummer: MFCD00866661
CAS-nummer: 146939-27-7
Merck Indeks: 14,10171

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 218.0 to 224.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR