Zinc meso-tetraphenylporphine ≥97.0% (by HPLC)

Leverandør: TCI

Synonymer: 5,10,15,20-Tetraphenyl-21H,23H-porphine Zinc(II), Zinc(II) Tetraphenylporphyrin, 5,10,15,20-Tetraphenyl-21H,23H-porphine zinc, Zinc TPP, Zinc(II) TPP, Tetraphenylporphine zinc, TPP Zn

Z0036-1G Z0036-5G
TCIAZ0036-1GEA 1875 NOK
TCIAZ0036-1G TCIAZ0036-5G
Zinc meso-tetraphenylporphine ≥97.0% (by HPLC)
Zinc meso-tetraphenylporphine
Formel: C₄₄H₂₈N₄Zn
MW: 678,10 g/mol
MDL nummer: MFCD00012155
CAS-nummer: 14074-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4cb6142934beff576a90383dac7ed9ee5e854af&vid=a4cb6142934beff576a90383dac7ed9ee5e854af&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Green to Dark green to Dark blue powder to crystal
Purity (HPLC) min. 97.0 area%
Content of Zinc 8.6 to 10.6 %
Lambda max. 420.0 to 426.0 nm (THF)
Molar extinction coefficient min. 625000 (THF, 420.0 to 426.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR