4'-Hydroxy-3'-methoxyacetophenone ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
A10439.18 A10439.30 A10439.36
A10439.18EA 750 NOK
A10439.18 A10439.30 A10439.36
4'-Hydroxy-3'-methoxyacetophenone ≥98%
4'-Hydroxy-3'-methoxyacetophenone
Purity: 98% CAS No.: 498-02-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 112-116° Boiling Point: 295-300° Merck Reference: 14,741 MDL No.: MFCD00008747
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokepunkt: 295…300 °C
Smeltepunkt: 112…116 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008747
CAS-nummer: 498-02-2
EINECS: 207-854-5
Merck Indeks: 14,741