Industrial gloves, Edge® 48-125
Leverandør: Ansell
48-125/8 48-125/7 48-125/6 48-125/10 48-125/11 48-125/9
ANSE48-125/8EA 310 NOK
ANSE48-125/8 ANSE48-125/7 ANSE48-125/6 ANSE48-125/10 ANSE48-125/11 ANSE48-125/9
Industrial gloves, Edge® 48-125
Hansker
These knitted polyester liners with PU dip coating provide a good level of abrasion resistance and grip, as well as high tactility in dry to slightly oily environments.
  • 15 gauge highly comfortable and dexterous polyester liner
  • Seamless knitted liner with elasticized knit wrist dipped into white polyurethane coating
  • Good flexibility and finger dexterity compared to cotton canvas or thick cotton gloves
  • White colour shows dirt contamination more quickly
Recommended for: Assembly of small pieces, general handling, packaging, quality inspection.