1-Hydroxy-2-naphthoic acid 98%
Leverandør: Thermo Scientific
Warning
Synonymer: 1-Hydroxy-naphthalene-2-carboxylic acid
149230050 149231000
ACRO149230050EA 320 NOK
ACRO149230050 ACRO149231000
1-Hydroxy-2-naphthoic acid 98%
1-Hydroxy-2-naphthoic acid
Formel: C₁₁H₈O₃
MW: 188,18 g/mol
Kokepunkt: 375 °C (1013 hPa)
Smeltepunkt: 199…205 °C
Flash punkt: >150 °C
MDL nummer: MFCD00003960
CAS-nummer: 86-48-6
EINECS: 201-674-0
ADR: 6.1,III
Merck Indeks: 13,04859
Bestill nå
Spesifikasjon Testresultater
Appearance Cream to beige to grey-brown Powder
Infrared spectrum Conforms
Melting point 195 to 205 °C (Dec)
Titration with NaOH ≥97.5 %
Specific optical rotation -36.0° to -40.0° (20 °C, 589 nm) (c=1 in dioxane)