2,4,6-Trichloro-1,3,5-triazine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
Cyanuric chloride, 2,4,6-Trichloro-s-triazine
C0460-25G
C0460-500G
TCIAC0460-25GEA
230
NOK
TCIAC0460-25G
TCIAC0460-500G
2,4,6-Trichloro-1,3,5-triazine ≥98.0% (by titrimetric analysis)
2,4,6-Trichloro-1,3,5-triazine
Formel:
C₃Cl₃N₃ MW: 184,41 g/mol Kokepunkt: 190 °C (1013 hPa) Tetthet: 1,32 g/cm³ (20 °C) |
MDL nummer:
MFCD00006046 CAS-nummer: 108-77-0 EINECS: 203-614-9 FN: 2670 ADR: 8,II Merck Indeks: 14,02699 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 145.0 to 150.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...