Industrial label printers, BradyPrinter i7100
Leverandør: Brady
149047 198613 149046
BRDY149047EA 37750 NOK
BRDY149047 BRDY198613 BRDY149046
Industrial label printers, BradyPrinter i7100
Printere Label Printers
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the Aerospace, Defence and Mass Transit, Electronics, Automotive, Logistics, Electrical, Voice-Datacom and LAB markets.
  • 300 or 600 dpi
  • Applications include cable and wire identification, circuit board and component identification, and general and industrial identification
  • Mono colour
  • Print speed upto 300 mm/s
  • Powerful internal processor designed to handle up to 7000 labels per day
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Minne 256 MB RAM, 50 MB IFFS Flash, SD card,
slot up to 512 GB, internal
Oppløsning 300 or 600 dpi
Dimensjoner 252W×288H×460D mm
Vekt 10 kg