Avantor® Hichrom HI-1, GC Columns
Leverandør: Avantor
HI01-05-005-2EA
5100
NOK
HI01-05-005-2
HI01-05-005-2.5
HI01-10-010-10
HI01-10-010-15
HI01-10-010-5
HI01-15-015-20
HI01-15-015-5
HI01-25-025-15G5
HI01-25-025-30G5
HI01-25-025-60G5
HI01-53-100-30G5
HI01-18-018-20
HI01-18-020-10
HI01-18-020-40
HI01-18-040-40
HI01-18-040-50
HI01-20-010-10
HI01-20-020-40
HI01-20-040-20
HI01-20-100-40
HI01-25-010-15
HI01-25-010-30
HI01-25-025-105
HI01-25-025-15
HI01-25-025-50
HI01-25-050-105
HI01-25-050-50
HI01-25-100-10
HI01-25-100-105
HI01-25-100-30
HI01-25-100-50
HI01-25-100-60
HI01-32-010-10
HI01-32-010-105
HI01-32-010-15
HI01-32-010-30
HI01-32-010-50
HI01-32-010-60
HI01-32-025-105
HI01-32-025-50
HI01-32-025-60
HI01-32-050-10
HI01-32-050-105
HI01-32-050-50
HI01-32-100-105
HI01-32-150-105
HI01-32-150-50
HI01-32-300-105
HI01-32-400-30
HI01-32-500-105
HI01-32-500-30
HI01-32-500-60
HI01-53-010-10
HI01-53-010-30
HI01-53-050-10
HI01-53-100-10
HI01-53-100-15
HI01-53-100-50
HI01-53-150-10
HI01-53-150-15
HI01-53-150-75
HI01-53-300-10
HI01-53-300-15
HI01-53-300-30
HI01-53-300-50
HI01-53-500-10
HI01-53-500-50
HI01-53-500-75
HIGCHI01-32-052-25
HIGCHI01-25-030-30
HIGCHI01-53-265-30
HIGCHI01-53-500-25
HIGCHI01-53-015-15
HIGCHI015300905MTX
HIGCHI015308810MTX
HIGCHI01-32-120-50
HIGCHI01-25-025-25
HIGCHI01-32-120-25
HIGCHI01-32-120-20
HIGCHI01-32-132-25
Avantor® Hichrom HI-1, GC Columns
Kromatografikolonner
SPESIFIKASJONER
Environmentally Preferable | |
Phase name | HI-1 |
Phase composition | 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) |
Polarity | Apolar stationary phase |
Crossbond | Yes |
Temperaturgrense | 350 °C |
USP kode | G1, G2, G9, G38 |