Avantor® Hichrom HI-1701, GC Columns
Leverandør: Avantor
HI06-05-005-2EA
5450
NOK
HI06-05-005-2
HI06-05-005-2.5
HI06-05-005-5
HI06-10-010-10
HI06-10-010-15
HI06-10-010-5
HI06-10-020-10
HI06-10-020-15
HI06-10-020-5
HI06-10-040-10
HI06-10-040-15
HI06-10-040-5
HI06-15-015-10
HI06-15-015-15
HI06-15-015-20
HI06-15-015-30
HI06-15-015-5
HI06-25-025-15G5
HI06-25-025-30G5
HI06-25-025-60G5
HI06-53-100-30G5
HI06-18-018-10
HI06-18-018-20
HI06-18-018-40
HI06-18-018-50
HI06-18-018-60
HI06-18-020-10
HI06-18-020-20
HI06-18-020-40
HI06-18-020-50
HI06-18-020-60
HI06-18-040-10
HI06-18-040-20
HI06-18-040-40
HI06-18-040-50
HI06-18-040-60
HI06-20-018-10
HI06-20-018-20
HI06-20-018-40
HI06-20-018-50
HI06-20-018-60
HI06-20-020-10
HI06-20-020-20
HI06-20-020-40
HI06-20-020-50
HI06-20-020-60
HI06-20-040-10
HI06-20-040-20
HI06-20-040-40
HI06-20-040-50
HI06-20-040-60
HI06-25-010-15
HI06-25-010-30
HI06-25-010-50
HI06-25-010-60
HI06-25-025-10
HI06-25-025-105
HI06-25-025-15
HI06-25-025-30
HI06-25-025-50
HI06-25-025-60
HI06-25-050-10
HI06-25-050-105
HI06-25-050-15
HI06-25-050-30
HI06-25-050-50
HI06-25-050-60
HI06-25-100-10
HI06-25-100-105
HI06-25-100-15
HI06-25-100-30
HI06-25-100-50
HI06-25-100-60
HI06-32-010-10
HI06-32-010-105
HI06-32-010-15
HI06-32-010-30
HI06-32-010-50
HI06-32-010-60
HI06-32-025-10
HI06-32-025-105
HI06-32-025-15
HI06-32-025-30
HI06-32-025-50
HI06-32-025-60
HI06-32-050-10
HI06-32-050-105
HI06-32-050-15
HI06-32-050-30
HI06-32-050-50
HI06-32-050-60
HI06-32-100-10
HI06-32-100-105
HI06-32-100-15
HI06-32-100-30
HI06-32-100-50
HI06-32-100-60
HI06-32-150-10
HI06-32-150-105
HI06-32-150-15
HI06-32-150-30
HI06-32-150-50
HI06-32-150-60
HI06-32-300-10
HI06-32-300-105
HI06-32-300-15
HI06-32-300-30
HI06-32-300-50
HI06-32-300-60
HI06-53-010-10
HI06-53-010-15
HI06-53-010-30
HI06-53-025-15
HI06-53-025-30
HI06-53-025-50
HI06-53-025-60
HI06-53-050-10
HI06-53-050-15
HI06-53-050-30
HI06-53-050-50
HI06-53-050-60
HI06-53-100-10
HI06-53-100-15
HI06-53-100-30
HI06-53-100-50
HI06-53-100-60
HI06-53-100-75
HI06-53-150-10
HI06-53-150-15
HI06-53-150-30
HI06-53-150-50
HI06-53-150-60
HI06-53-150-75
HI06-53-300-10
HI06-53-300-15
HI06-53-300-30
HI06-53-300-50
HI06-53-300-60
HI06-53-300-75
HIGCHI06-25-030-30
HIGCHI06-32-050-25
Avantor® Hichrom HI-1701, GC Columns
Kromatografikolonner
SPESIFIKASJONER
Environmentally Preferable | |
Phase name | HI-1701 |
Phase composition | 14% Cyanopropylphenyl, 86% Methyl Polysiloxane |
Polarity | Mid-polarity stationary phase |
Crossbond | Yes |
Temperaturgrense | 280 °C |
USP kode | G46 |