Avantor® Hichrom HI-17, GC Columns
Leverandør: Avantor
HI07-05-005-2EA 5450 NOK
HI07-05-005-2 HI07-05-005-2.5 HI07-05-005-5 HI07-10-010-10 HI07-10-010-15 HI07-10-010-5 HI07-10-020-10 HI07-10-020-15 HI07-10-020-5 HI07-10-040-10 HI07-10-040-15 HI07-10-040-5 HI07-15-015-10 HI07-15-015-15 HI07-15-015-20 HI07-15-015-30 HI07-15-015-5 HI07-25-025-15G5 HI07-25-025-30G5 HI07-25-025-60G5 HI07-53-100-30G5 HI07-18-018-10 HI07-18-018-20 HI07-18-018-40 HI07-18-018-50 HI07-18-018-60 HI07-18-020-10 HI07-18-020-20 HI07-18-020-40 HI07-18-020-50 HI07-18-020-60 HI07-18-040-10 HI07-18-040-20 HI07-18-040-40 HI07-18-040-50 HI07-18-040-60 HI07-20-010-10 HI07-20-010-20 HI07-20-010-40 HI07-20-010-50 HI07-20-010-60 HI07-20-020-10 HI07-20-020-20 HI07-20-020-40 HI07-20-020-50 HI07-20-020-60 HI07-20-040-10 HI07-20-040-20 HI07-20-040-40 HI07-20-040-50 HI07-20-040-60 HI07-20-100-10 HI07-20-100-20 HI07-20-100-40 HI07-20-100-60 HI07-25-010-15 HI07-25-010-30 HI07-25-010-50 HI07-25-010-60 HI07-25-025-10 HI07-25-025-105 HI07-25-025-15 HI07-25-025-30 HI07-25-025-50 HI07-25-025-60 HI07-25-050-10 HI07-25-050-105 HI07-25-050-15 HI07-25-050-30 HI07-25-050-50 HI07-25-050-60 HI07-25-100-10 HI07-25-100-105 HI07-25-100-15 HI07-25-100-30 HI07-25-100-50 HI07-25-100-60 HI07-32-010-10 HI07-32-010-105 HI07-32-010-15 HI07-32-010-30 HI07-32-010-50 HI07-32-010-60 HI07-32-025-10 HI07-32-025-105 HI07-32-025-15 HI07-32-025-30 HI07-32-025-50 HI07-32-025-60 HI07-32-050-10 HI07-32-050-105 HI07-32-050-15 HI07-32-050-30 HI07-32-050-50 HI07-32-050-60 HI07-32-100-10 HI07-32-100-105 HI07-32-100-15 HI07-32-100-30 HI07-32-100-50 HI07-32-100-60 HI07-32-150-10 HI07-32-150-105 HI07-32-150-15 HI07-32-150-30 HI07-32-150-50 HI07-32-150-60 HI07-32-300-10 HI07-32-300-105 HI07-32-300-15 HI07-32-300-30 HI07-32-300-50 HI07-32-300-60 HI07-53-010-10 HI07-53-010-15 HI07-53-010-30 HI07-53-025-15 HI07-53-025-30 HI07-53-025-50 HI07-53-025-60 HI07-53-050-10 HI07-53-050-15 HI07-53-050-30 HI07-53-050-50 HI07-53-050-60 HI07-53-100-10 HI07-53-100-15 HI07-53-100-30 HI07-53-100-50 HI07-53-100-60 HI07-53-100-75 HI07-53-150-10 HI07-53-150-15 HI07-53-150-30 HI07-53-150-50 HI07-53-150-60 HI07-53-150-75 HI07-53-265-10 HI07-53-265-30 HI07-53-300-10 HI07-53-300-15 HI07-53-300-30 HI07-53-300-50 HI07-53-300-60 HI07-53-300-75 HIGCHI07-32-050-25 HIGCHI07-25-015-30 HIGCHI075310030/5
Avantor® Hichrom HI-17, GC Columns
Kromatografikolonner
General purpose polar phase for a wide range of applications, ideal as confirmation analysis column. Ideal in a GCxGC configuration as polar dimension column. HI-17 is a Crossbonded phase and assure a long lifetime even under high temperature limits.
  • Phase composition: crossbonded 50% Phenyl, 50% Methyl Polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.
Bestill nå
SPESIFIKASJONER
Phase name HI-17
Environmentally Preferable
Phase composition 50% Phenyl, 50% Methyl Polysiloxane
Polarity Mid-to-high polarity stationary phase
Crossbond Stabilised
Temperaturgrense 340 °C
USP kode G3, G17