Avantor® Hichrom HI-624 MS, GC Columns
Leverandør: Avantor
HI24-25-025-15G5EA 7325 NOK
HI24-25-025-15G5 HI24-25-025-30G5 HI24-25-025-60G5 HI24-25-140-30G5 HI24-25-140-60G5 HI24-32-180-30G5 HI24-53-100-30G5 HI24-53-300-30G5 HI24-18-018-10 HI24-18-018-20 HI24-18-018-40 HI24-18-018-50 HI24-18-018-60 HI24-18-020-10 HI24-18-020-20 HI24-18-020-40 HI24-18-020-50 HI24-18-020-60 HI24-18-040-10 HI24-18-040-20 HI24-18-040-40 HI24-18-040-50 HI24-18-100-20 HI24-18-100-40 HI24-18-100-50 HI24-18-100-60 HI24-20-112-20 HI24-20-112-40 HI24-20-112-50 HI24-20-112-60 HI24-25-010-15 HI24-25-010-30 HI24-25-010-50 HI24-25-010-60 HI24-25-025-10 HI24-25-025-105 HI24-25-025-15 HI24-25-025-30 HI24-25-025-50 HI24-25-025-60 HI24-25-050-10 HI24-25-050-105 HI24-25-050-15 HI24-25-050-30 HI24-25-050-50 HI24-25-050-60 HI24-25-100-10 HI24-25-100-15 HI24-25-100-30 HI24-25-100-50 HI24-25-100-60 HI24-25-140-105 HI24-25-140-30 HI24-25-140-50 HI24-25-140-60 HI24-25-300-30 HI24-25-300-50 HI24-25-300-60 HI24-32-010-10 HI24-32-010-105 HI24-32-010-15 HI24-32-010-30 HI24-32-010-50 HI24-32-010-60 HI24-32-025-10 HI24-32-025-105 HI24-32-025-15 HI24-32-025-30 HI24-32-025-50 HI24-32-025-60 HI24-32-050-10 HI24-32-050-105 HI24-32-050-15 HI24-32-050-30 HI24-32-050-50 HI24-32-050-60 HI24-32-100-10 HI24-32-100-105 HI24-32-100-15 HI24-32-100-30 HI24-32-100-50 HI24-32-100-60 HI24-32-150-10 HI24-32-150-105 HI24-32-150-15 HI24-32-150-30 HI24-32-150-50 HI24-32-150-60 HI24-32-180-105 HI24-32-180-30 HI24-32-180-50 HI24-32-180-60 HI24-32-180-75 HI24-32-300-10 HI24-32-300-105 HI24-32-300-15 HI24-32-300-30 HI24-32-300-50 HI24-32-300-60 HI24-53-010-10 HI24-53-010-15 HI24-53-010-30 HI24-53-025-15 HI24-53-025-30 HI24-53-025-50 HI24-53-025-60 HI24-53-050-10 HI24-53-050-15 HI24-53-050-30 HI24-53-050-50 HI24-53-050-60 HI24-53-100-10 HI24-53-100-15 HI24-53-100-30 HI24-53-100-50 HI24-53-100-60 HI24-53-100-75 HI24-53-150-10 HI24-53-150-15 HI24-53-150-30 HI24-53-150-50 HI24-53-150-60 HI24-53-150-75 HI24-53-300-10 HI24-53-300-15 HI24-53-300-30 HI24-53-300-50 HI24-53-300-60 HI24-53-300-75 HI24-05-005-2 HI24-05-005-2.5 HI24-05-005-5 HI24-10-010-10 HI24-10-010-15 HI24-10-010-5 HI24-10-020-10 HI24-10-020-15 HI24-10-020-5 HI24-10-040-10 HI24-10-040-15 HI24-10-040-5 HI24-15-015-10 HI24-15-015-15 HI24-15-015-20 HI24-15-015-30 HI24-15-015-5 HI24-15-084-15 HI24-15-084-20 HI24-15-084-30 HI24-15-084-40
Avantor® Hichrom HI-624 MS, GC Columns
Kromatografikolonner
General purpose low to mid polarity phase for a wide range of applications, including (for example): Residual solvents, volatile pollutants. HI-624 MS is an MS grade cross-bonded column.
  • Phase composition: Cross-bonded 6% Cyanopropyl phenyl, 94% Methyl Polysiloxane - MS grade
  • Long lifetime
Max. temperature limits may change depending on the film thickness.