Avantor® Hichrom HI-225, GC Columns
Leverandør: Avantor
HI33-18-018-10EA 5925 NOK
HI33-18-018-10 HI33-18-018-20 HI33-18-018-40 HI33-18-018-50 HI33-18-018-60 HI33-18-020-10 HI33-18-020-20 HI33-18-020-40 HI33-18-040-10 HI33-18-040-20 HI33-18-040-40 HI33-18-040-50 HI33-18-040-60 HI33-20-010-10 HI33-20-010-20 HI33-20-010-40 HI33-20-010-50 HI33-20-010-60 HI33-20-020-10 HI33-20-020-20 HI33-20-020-40 HI33-20-020-50 HI33-20-020-60 HI33-20-040-10 HI33-20-040-20 HI33-20-040-40 HI33-20-040-50 HI33-20-040-60 HI33-20-100-10 HI33-20-100-20 HI33-20-100-40 HI33-20-100-50 HI33-20-100-60 HI33-25-010-15 HI33-25-010-30 HI33-25-010-50 HI33-25-010-60 HI33-25-025-10 HI33-25-025-105 HI33-25-025-15 HI33-25-025-30 HI33-25-025-50 HI33-25-025-60 HI33-25-050-10 HI33-25-050-105 HI33-25-050-15 HI33-25-050-30 HI33-25-050-50 HI33-25-050-60 HI33-32-010-10 HI33-32-010-105 HI33-32-010-15 HI33-32-010-30 HI33-32-010-50 HI33-32-010-60 HI33-32-025-10 HI33-32-025-105 HI33-32-025-15 HI33-32-025-30 HI33-32-025-50 HI33-32-025-60 HI33-32-050-10 HI33-32-050-105 HI33-32-050-15 HI33-32-050-30 HI33-32-050-50 HI33-32-050-60 HI33-32-100-10 HI33-32-100-105 HI33-32-100-15 HI33-32-100-30 HI33-32-100-50 HI33-32-100-60 HI33-53-010-10 HI33-53-010-15 HI33-53-010-30 HI33-53-025-15 HI33-53-025-30 HI33-53-025-50 HI33-53-025-60 HI33-53-050-10 HI33-53-050-15 HI33-53-050-30 HI33-53-050-50 HI33-53-050-60 HI33-05-005-2 HI33-05-005-2.5 HI33-05-005-5 HI33-10-010-10 HI33-10-010-15 HI33-10-010-5 HI33-10-020-10 HI33-10-020-15 HI33-10-020-5 HI33-10-040-10 HI33-10-040-15 HI33-10-040-5 HI33-15-015-10 HI33-15-015-15 HI33-15-015-20 HI33-15-015-30 HI33-15-015-5
Avantor® Hichrom HI-225, GC Columns
Kromatografikolonner
General purpose mid-to-high-polarity phase for a wide range of applications, like, for example: FAMEs, carbohydrates, sterols, flavor compounds.
  • Phase composition: Crossbonded 25% Cyanopropyl, 25% Phenyl, 50% Methyl Polysiloxane
Max temperature limits may change depending on the film thickness.
Bestill nå
SPESIFIKASJONER
Phase name HI-225
Environmentally Preferable
Phase composition 25% Cyanopropyl, 25% Phenyl, 50% Methyl Polysiloxane
Polarity Mid-to-high polarity stationary phase
Crossbond Stabilised
Temperaturgrense 260 °C
USP kode G7, G19, G26