Avantor® Hichrom HI-ALC 2, GC Columns
Leverandør: Avantor
HI37-32-060-30EA 12300 NOK
HI37-32-060-30 HI37-32-100-30 HI37-53-100-30
Avantor® Hichrom HI-ALC 2, GC Columns
Kromatografikolonner
Application specific column for blood alcohols testing.
Max temperature limits may change depending on the film thickness.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Phase name HI-ALC 2
Phase composition Proprietary Phase developed for blood alcohol analysis
Crossbond Yes
Temperaturgrense 280 °C