Avantor® Hichrom HI-1 PONA, GC Columns
Leverandør: Avantor
HI57-20-050-100EA 29025 NOK
HI57-20-050-100 HI57-20-050-50 HI57-25-050-100 HI57-25-100-150
Avantor® Hichrom HI-1 PONA, GC Columns
Kromatografikolonner
Optimized phase for DHA (Detailed Hydrocarbons Analysis), PONA, PIANO and PNA analysis.
Max temperature limits may change depending on the film thickness.
Bestill nå
SPESIFIKASJONER
Phase name HI-1 PONA
Environmentally Preferable
Phase composition 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - optimized for hydrocarbon analysis
Polarity Apolar stationary phase
Crossbond Yes
Temperaturgrense 350 °C