Avantor® Hichrom HI-DEX DAC Gamma, GC Columns
Leverandør: Avantor
HI67-25-015-15EA 10225 NOK
HI67-25-015-15 HI67-25-015-25 HI67-25-015-30 HI67-25-015-50 HI67-25-015-60 HI67-25-025-15 HI67-25-025-25 HI67-25-025-30 HI67-25-025-50 HI67-25-025-60 HI79-10-010-10 HI79-10-010-5
Avantor® Hichrom HI-DEX DAC Gamma, GC Columns
Kromatografikolonner
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Bestill nå
SPESIFIKASJONER
Phase name HI-DEX DAC Gamma
Environmentally Preferable
Phase composition Diacethyl TBS Gamma Cyclodexstrin based
Polarity Chiral stationary phase
Crossbond No
Temperaturgrense 230 °C