Avantor® Hichrom HI-35 HT, GC Columns
Leverandør: Avantor
HI89-25-010-15EA 6775 NOK
HI89-25-010-15 HI89-25-010-30 HI89-25-015-15 HI89-25-015-30 HI89-25-025-15 HI89-25-025-30 HI89-32-010-10 HI89-32-010-15 HI89-32-010-30 HI89-32-015-15 HI89-32-015-30 HI89-32-025-15 HI89-32-025-30 HI89-53-010-10 HI89-53-010-15 HI89-53-010-30 HI89-53-015-15 HI89-53-015-30 HI89-53-017-30 HI89-53-025-15 HI89-53-025-30 HI89-10-010-10 HI89-10-010-5
Avantor® Hichrom HI-35 HT, GC Columns
Kromatografikolonner
General purpose column with added high temperature stability for the analysis of Pesticides, PCBs, substituted polar compounds, phenols, etc. Ideal as confrmation column.
Max. temperature limits may change depending on the film thickness.
Bestill nå
SPESIFIKASJONER
Phase name HI-35 HT
Environmentally Preferable
Phase composition 35% Phenyl, 65% Methyl Polysiloxane - high temperature
Polarity Mid-polarity stationary phase
Crossbond Stabilised
Temperaturgrense 370 °C
USP kode G28, G32, G42