Pipetter, serologisk, PYREX®
Leverandør: Corning
7058-2X 7077-10N 7077-1N 7077-2N 7077-5N 7077B-1 7077B-10 7077B-25 7077B-5 7077B-50 7078-10N 7078-1CN 7078-1N 7078-2N 7078-5N 7078B-1 7078B-10 7078B-25 7078B-5 7078B-50 7079-10N 7079-1N 7079-2N 7079-5N 7078D-1 7078D-10 7078D-5
612-5130EA 4450 NOK
612-5130 612-5131 612-5132 612-5133 612-5134 612-5136 612-5138 612-5139 612-5140 612-5141 612-5162 612-5163 612-5164 612-5165 612-5166 612-5177 612-5178 612-5186 612-5187 612-5188 612-5189 612-5192 612-5193 612-5194 612-5748 612-5749 612-5750
Pipetter, serologisk, PYREX®
Pipetter Serologiske pipetter
These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.
  • All pipettes have negative graduations
  • Fire-polished tips for burr-free, even flow rates
Easy-open Steri-View™ wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size.
Sertifiseringer: Colors conform to I.S.O. standards.
Forpakning: Non sterile pipettes available in bulk package; sterile pipettes available inMore Product Information