VWR® ELSD 90 LT, ELSD 85 LT, ELSD 80 LT, Low Temperature Evaporative Light-Scattering Detectors
Leverandør: VWR Collection
903-0245EA 0 NOK
903-0245 903-0234 903-0250 903-0249 903-0248 903-0244 903-0243 903-0242 903-0241 903-0240 903-0239 903-0238 903-0247 903-0235 903-0236 903-0237 903-0271 903-0272 903-0273 903-0274
VWR® ELSD 90 LT, ELSD 85 LT, ELSD 80 LT, Low Temperature Evaporative Light-Scattering Detectors
Kromatografidetektorer
These detectors include a number of innovative features including the ability to select the best nebuliser and a unique low temperature technology. The evaporation drift-tube design optimises both resolution and sensitivity. In addition, you can control the system locally or via a PC. A remote shutdown mode is provided to minimise cost and enhance system lifetime.
  • Complete remote control
  • Sensitivity is enhanced by digital signal treatment
  • Full SOP protocols are provided for GLP compliance and validation procedures
These systems allow you to detectMore Product Information