Gloves, neoprene-nitrile, multi-layered, Kimtech™ Prizm™
Leverandør: KIMBERLY CLARK
99221 99223
KIMB99221EA 0 NOK
KIMB99221 112-6130
Gloves, neoprene-nitrile, multi-layered, Kimtech™ Prizm™
Hansker
Kimtech™ Prizm™ multi-layered neoprene nitrile gloves, designed for scientists by scientists, are an ideal choice for the most demanding scientific applications. One of the thinnest Type A certified protective gloves, providing the perfect balance between precision, protection, and comfort.
  • Protection against a wide range of chemicals
  • Ultra-fingertip grip, even when wet
  • Certified ergonomic comfort
  • Mechanical breach indicator
  • Latex-free, powder-free safety gloves with a nitrile accelerator-free donning layer
Seamlessly protect people and processes whenMore Product Information
Bestill nå
SPESIFIKASJONER
Ambidekstral/håndspesifikk Ambidextrous
Engangs/flergangs engangs
Latex/latexfri Latex-fri
Pudder/pudderfri uten pudder
Overflate Textured fingertips
Type mansjett Perlebesatt
Tykkelse mansjett 0,11 mm
Tykkelse finger 0,22 mm
Mansjett tykkelse 0,16 mm
Tensile strength 14 MPa, nominal
Ultimat elongering 400% nominal (after accelerated aging)
EN norm EN ISO 374-1:2016 Type A (JKLMPT), EN ISO 374-5:2016 Micro-Organism/VIRUS