5,7-Dichloro-8-hydroxyquinoline sulphate, sodium salt ≥95%
Leverandør: Apollo Scientific
Warning
Synonymer: 5,7-Dichloro-8-(hydrogen sulphate)-8-quinolinol sodium salt
BICL4299-10MG BICL4299-5MG
APOSBICL4299-10MGEA 12050 NOK
APOSBICL4299-10MG APOSBICL4299-5MG
5,7-Dichloro-8-hydroxyquinoline sulphate, sodium salt ≥95%
5,7-Dichloro-8-hydroxyquinoline sulphate, sodium salt
Smeltepunkt: 240…245 °C
Lagringstemperatur: Fryseskap
CAS-nummer: 58698-96-7