3-Bromo-2-phenylimidazo[1,2-b]pyridazine

Leverandør: Apollo Scientific
Warning

Synonymer: (3-Bromoimidazo[1,2-b]pyridazin-2-yl)benzene

OR01787-1G OR01787-500MG
APOSOR01787-1GEA 4825 NOK
APOSOR01787-1G APOSOR01787-500MG
3-Bromo-2-phenylimidazo[1,2-b]pyridazine
3-Bromo-2-phenylimidazo[1,2-b]pyridazine
Formel: C₁₂H₈BrN₃
MW: 274,116 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09832525
CAS-nummer: 802580-12-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR