2-Methyl-4-(piperazin-1-yl)quinoline

Leverandør: Apollo Scientific
Danger

Synonymer: 1-(2-Methylquinolin-4-yl)piperazine

OR27760-250MG OR27760-1G
APOSOR27760-250MGEA 1450 NOK
APOSOR27760-250MG APOSOR27760-1G
2-Methyl-4-(piperazin-1-yl)quinoline
2-Methyl-4-(piperazin-1-yl)quinoline
Formel: C₁₄H₁₇N₃
MW: 227,305 g/mol
Smeltepunkt: 100…101 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01935745
CAS-nummer: 82241-22-3
FN: 3259
ADR: 8,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR