6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroquinolin-2(1H)-one

Leverandør: Apollo Scientific
Warning

OR300010-1G OR300010-250MG
APOSOR300010-1GEA 2775 NOK
APOSOR300010-1G APOSOR300010-250MG
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroquinolin-2(1H)-one
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroquinolin-2(1H)-one
Formel: C₁₅H₂₀BNO₃
MW: 273,135 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18072003
CAS-nummer: 400620-72-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR