3,4-Dihydro-2H-pyran-5-carboxylic acid 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 5-Carboxy-3,4-dihydro-2H-pyran

OR303311-25G
APOSOR303311-25GEA 1050 NOK
APOSOR303311-25G
3,4-Dihydro-2H-pyran-5-carboxylic acid 98%
3,4-Dihydro-2H-pyran-5-carboxylic acid
Formel: C₆H₈O₃
MW: 128,126 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06203171
CAS-nummer: 40915-37-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR