2-Iodo-1H-indole
Leverandør: Apollo Scientific
|
Warning
|
OR305199-1G
APOSOR305199-1GEA
4050
NOK
APOSOR305199-1G
2-Iodo-1H-indole
2-Iodo-1H-indole
Formel:
C₈H₆IN MW: 243,044 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD10699391 CAS-nummer: 26340-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...