2-Amino-4-ethylphenol hydrochloride

Leverandør: Apollo Scientific
Warning

OR451457-1G OR451457-5G
APOSOR451457-1GEA 2150 NOK
APOSOR451457-1G APOSOR451457-5G
2-Amino-4-ethylphenol hydrochloride
2-Amino-4-ethylphenol hydrochloride
Formel: C₈H₁₁NO·ClH
MW: 173,64 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD21602524
CAS-nummer: 79509-90-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR