1-Benzhydryl-3-iodo-azetidine 98%
Leverandør: Apollo Scientific
Synonymer:
1-Benzhydryl-3-iodoazetidine
OR480620-1G
OR480620-5G
APOSOR480620-1GEA
1200
NOK
APOSOR480620-1G
APOSOR480620-5G
1-Benzhydryl-3-iodo-azetidine 98%
1-Benzhydryl-3-iodo-azetidine
Formel:
C₁₆H₁₆IN MW: 349,209 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD08062427 CAS-nummer: 125735-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...