2-Chloro-7,8-dihydroquinolin-5(6H)-one 97%

Leverandør: Apollo Scientific

OR54586-5G
APOSOR54586-5GEA 1575 NOK
APOSOR54586-5G
2-Chloro-7,8-dihydroquinolin-5(6H)-one 97%
2-Chloro-7,8-dihydroquinolin-5(6H)-one
Formel: C₉H₈ClNO
MW: 181,619 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08692066
CAS-nummer: 124467-36-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR