5-Chloro-2-iodophenol
Leverandør: Apollo Scientific
|
Warning
|
OR70040-1G
OR70040-5G
APOSOR70040-1GEA
645
NOK
APOSOR70040-1G
APOSOR70040-5G
5-Chloro-2-iodophenol
5-Chloro-2-iodophenol
Formel:
C₆H₄ClIO MW: 254,453 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD09753473 CAS-nummer: 136808-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...