4-(Dimethoxymethyl)benzyl alcohol 98%

Leverandør: Apollo Scientific

OR900743-10G OR900743-1G OR900743-5G
APOSOR900743-10GEA 6625 NOK
APOSOR900743-10G APOSOR900743-1G APOSOR900743-5G
4-(Dimethoxymethyl)benzyl alcohol 98%
4-(Dimethoxymethyl)benzyl alcohol
Formel: C₁₀H₁₄O₃
MW: 182,216 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01863763
CAS-nummer: 183057-64-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR