1-(2-Methylphenyl)-1H-pyrrole-2-carbaldehyde 95%

Leverandør: Apollo Scientific

OR905190-1G OR905190-5G
APOSOR905190-1GEA 2425 NOK
APOSOR905190-1G APOSOR905190-5G
1-(2-Methylphenyl)-1H-pyrrole-2-carbaldehyde 95%
1-(2-Methylphenyl)-1H-pyrrole-2-carbaldehyde
Formel: C₁₂H₁₁NO
MW: 185,222 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02664908
CAS-nummer: 35524-41-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR