1-Butyl-4-nitropyrazole 98%
Leverandør: Apollo Scientific
OR911094-1G
OR911094-25G
OR911094-5G
APOSOR911094-1GEA
1800
NOK
APOSOR911094-1G
APOSOR911094-25G
APOSOR911094-5G
1-Butyl-4-nitropyrazole 98%
1-Butyl-4-nitropyrazole
Formel:
C₇H₁₁N₃O₂ MW: 169,181 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD16810712 CAS-nummer: 1240574-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...