3-(Ethylamino)-4-nitrophenol 97%
Leverandør: Apollo Scientific
OR913948-10G
OR913948-1G
OR913948-5G
APOSOR913948-10GEA
9300
NOK
APOSOR913948-10G
APOSOR913948-1G
APOSOR913948-5G
3-(Ethylamino)-4-nitrophenol 97%
3-(Ethylamino)-4-nitrophenol
Formel:
C₈H₁₀N₂O₃ MW: 182,177 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD25121745 CAS-nummer: 25186-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...