4-[2-(Dimethylamino)ethyl]aniline 97%

Leverandør: Apollo Scientific

OR918366-10G OR918366-1G OR918366-5G
APOSOR918366-10GEA 3725 NOK
APOSOR918366-10G APOSOR918366-1G APOSOR918366-5G
4-[2-(Dimethylamino)ethyl]aniline 97%
4-[2-(Dimethylamino)ethyl]aniline
Formel: C₁₀H₁₆N₂
MW: 164,247 g/mol
Kokepunkt: 137…138 °C (6 mm Hg)
Smeltepunkt: 40…42 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04114509
CAS-nummer: 5636-52-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR