3-Chloro-4-nitro-1H-indole 95%
Leverandør: Apollo Scientific
OR919520-1G
OR919520-5G
APOSOR919520-1GEA
2250
NOK
APOSOR919520-1G
APOSOR919520-5G
3-Chloro-4-nitro-1H-indole 95%
3-Chloro-4-nitro-1H-indole
Formel:
C₈H₅ClN₂O₂ MW: 196,591 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD22689792 CAS-nummer: 208511-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...