2-(2-Furyl)-5-pyrimidinecarbaldehyde 95%

Leverandør: Apollo Scientific

Synonymer: 2-(Furan-2-yl)pyrimidine-5-carbaldehyde

OR924581-1G OR924581-5G
APOSOR924581-1GEA 2075 NOK
APOSOR924581-1G APOSOR924581-5G
2-(2-Furyl)-5-pyrimidinecarbaldehyde 95%
2-(2-Furyl)-5-pyrimidinecarbaldehyde
Formel: C₉H₆N₂O₂
MW: 174,156 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09864504
CAS-nummer: 959240-19-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR