Pyridine-4-sulphonyl chloride hydrochloride 98%
Leverandør: Apollo Scientific
|
Danger
|
OR926874-100MG
OR926874-250MG
OR926874-1G
OR926874-500MG
OR926874-2.5G
OR926874-5G
APOSOR926874-100MGEA
0
NOK
APOSOR926874-100MG
APOSOR926874-250MG
APOSOR926874-1G
APOSOR926874-500MG
APOSOR926874-2.5G
APOSOR926874-5G
Pyridine-4-sulphonyl chloride hydrochloride 98%
Pyridine-4-sulphonyl chloride hydrochloride
Formel:
C₅H₄ClNO₂S·ClH MW: 214,07 g/mol Smeltepunkt: 90…100 °C Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD04973456 CAS-nummer: 489430-50-4 FN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...